Anti-Hemoglobin alpha 1 Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB10772-100 ORB10772-200
BIRBORB10772-100EA 0 EUR
BIRBORB10772-100 BIRBORB10772-200
Anti-Hemoglobin alpha 1 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: Hemoglobin alpha 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human