Anti-SELL Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB12108-100 ORB12108-50
BIRBORB12108-100EA 0 EUR
BIRBORB12108-100 BIRBORB12108-50
Anti-SELL Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
FACS: 1:50-100

Type: Primary
Antigen: SELL (selectin L)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR