Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB12217-100 ORB12217-500
BIRBORB12217-100EA 0 EUR
BIRBORB12217-100 BIRBORB12217-500
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR