Anti-HTR2C Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB13225-100 ORB13225-20 ORB13225-5 ORB13225-50
BIRBORB13225-100EA 0 EUR
BIRBORB13225-100 BIRBORB13225-20 BIRBORB13225-5 BIRBORB13225-50
Anti-HTR2C Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: HTR2C (5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR