Anti-MAPK3 Goat Polyclonal Antibody

Supplier: Biorbyt
ORB18996-100 ORB18996-20
BIRBORB18996-100EA 0 EUR
BIRBORB18996-100 BIRBORB18996-20
Anti-MAPK3 Goat Polyclonal Antibody
Antibodies
ELISA: 1:4000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: MAPK3 (mitogen-activated protein kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR