Anti-Plasminogen Activator Goat Polyclonal Antibody

Supplier: Biorbyt
ORB19122-100 ORB19122-20
BIRBORB19122-100EA 0 EUR
BIRBORB19122-100 BIRBORB19122-20
Anti-Plasminogen Activator Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 0.3-1.0 ?g/ml

Type: Primary
Antigen: Plasminogen Activator
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR