Anti-WHSC1L1 Mouse Monoclonal Antibody
Supplier: Biorbyt
ORB22798-100 ORB22798-20 ORB22798-5
BIRBORB22798-100EA 0 EUR
BIRBORB22798-100 BIRBORB22798-20 BIRBORB22798-5
Anti-WHSC1L1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: WHSC1L1 (Wolf-Hirschhorn syndrome candidate 1-like 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human