Anti-ATG13 Rabbit polyclonal antibody
Supplier: Biorbyt
ORB48922-100 ORB48922-200
BIRBORB48922-100EA 0 EUR
BIRBORB48922-100 BIRBORB48922-200
Anti-ATG13 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: ATG13 (autophagy related 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat