Anti-ATG7 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB48923-100 ORB48923-200
BIRBORB48923-100EA 0 EUR
BIRBORB48923-100 BIRBORB48923-200
Anti-ATG7 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: ATG7 (autophagy related 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR