Anti-PPP3CA Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48981-100 ORB48981-200
BIRBORB48981-100EA 0 EUR
BIRBORB48981-100 BIRBORB48981-200
Anti-PPP3CA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP3CA (protein phosphatase 3, catalytic subunit, alpha isozyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR