Anti-SIK1 Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB100535-100 ORB100535-200
BIRBORB100535-100EA 0 EUR
BIRBORB100535-100 BIRBORB100535-200
Anti-SIK1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: SIK1 (salt-inducible kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now