Anti-NOC2L Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB101573-100 ORB101573-200
BIRBORB101573-100EA 0 EUR
BIRBORB101573-100 BIRBORB101573-200
Anti-NOC2L Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: NOC2L (nucleolar complex associated 2 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR