Anti-GTPase Activating Protein Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB106281-100 ORB106281-20
BIRBORB106281-100EA 0 EUR
BIRBORB106281-100 BIRBORB106281-20
Anti-GTPase Activating Protein Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: GTPase Activating Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR