Anti-IKBKG Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB106289-100 ORB106289-20
BIRBORB106289-100EA 0 EUR
BIRBORB106289-100 BIRBORB106289-20
Anti-IKBKG Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR