Anti-IRF3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125897-100 ORB125897-200
BIRBORB125897-100EA 0 EUR
BIRBORB125897-100 BIRBORB125897-200
Anti-IRF3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRF3 (interferon regulatory factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR