Anti-RUNX3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126324-100 ORB126324-200
BIRBORB126324-100EA 0 EUR
BIRBORB126324-100 BIRBORB126324-200
Anti-RUNX3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RUNX3 (runt-related transcription factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR