Anti-DKK3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126871-100 ORB126871-200
BIRBORB126871-100EA 0 EUR
BIRBORB126871-100 BIRBORB126871-200
Anti-DKK3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DKK3 (dickkopf WNT signaling pathway inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR