Anti-KCND1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127005-100 ORB127005-200
BIRBORB127005-100EA 0 EUR
BIRBORB127005-100 BIRBORB127005-200
Anti-KCND1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KCND1 (potassium voltage-gated channel, Shal-related subfamily, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR