Anti-PSMC6 Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB127167-100 ORB127167-200
BIRBORB127167-100EA 0 EUR
BIRBORB127167-100 BIRBORB127167-200
Anti-PSMC6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase, 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now