Anti-FOXO1/3/4-pan Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB127586-100 ORB127586-200
BIRBORB127586-100EA 0 EUR
BIRBORB127586-100 BIRBORB127586-200
Anti-FOXO1/3/4-pan Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FOXO1/3/4-pan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat