Anti-DLC1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127975-100 ORB127975-200
BIRBORB127975-100EA 0 EUR
BIRBORB127975-100 BIRBORB127975-200
Anti-DLC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DLC1 (deleted in liver cancer 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR