Anti-PARP1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB129005-100 ORB129005-200
BIRBORB129005-100EA 0 EUR
BIRBORB129005-100 BIRBORB129005-200
Anti-PARP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR