Anti-BIN1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB135048-100 ORB135048-200 ORB135048-50
BIRBORB135048-100EA 0 EUR
BIRBORB135048-100 BIRBORB135048-200 BIRBORB135048-50
Anti-BIN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BIN1 (bridging integrator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR