Anti-PAWR Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB136044-100 ORB136044-200 ORB136044-50
BIRBORB136044-100EA 0 EUR
BIRBORB136044-100 BIRBORB136044-200 BIRBORB136044-50
Anti-PAWR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PAWR (PRKC, apoptosis, WT1, regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR