Anti-AURKA Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB69374-10 ORB69374-100
BIRBORB69374-10EA 0 EUR
BIRBORB69374-10 BIRBORB69374-100
Anti-AURKA Mouse Monoclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400; ELISA: 1:10000

Type: Primary
Antigen: AURKA (aurora kinase A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR