Anti-CREB Rabbit polyclonal antibody unconjugated
Supplier: Biorbyt
ORB99340-100 ORB99340-20
BIRBORB99340-100EA 0 EUR
BIRBORB99340-100 BIRBORB99340-20
Anti-CREB Rabbit polyclonal antibody unconjugated
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500
Type: Primary
Antigen: CREB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat