Anti-CD23 Rabbit Monoclonal Antibody [clone: SP23]
Supplier: Abcam
AB16702-500 AB16702-1 AB16702-100
ABCAAB16702-500EA 725 EUR
ABCAAB16702-500 ABCAAB16702-1 ABCAAB16702-100
Anti-CD23 Rabbit Monoclonal Antibody [clone: SP23]
Antibodies
Rabbit monoclonal [SP23] to CD23.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: SP23
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human