Anti-Islet Cell Mouse Monoclonal Antibody [clone: LIV3D3]

Supplier: Abcam
AB22713-50 AB22713-100
ABCAAB22713-50EA 638 EUR
ABCAAB22713-50 ABCAAB22713-100
Anti-Islet Cell Mouse Monoclonal Antibody [clone: LIV3D3]
Antibodies
Mouse monoclonal [LIV3D3] to Islet Cell.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Islet Cell
Clonality: Monoclonal
Clone: LIV3D3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR