Anti-General Receptor for phosphoinositides 1/GRASP Rabbit Monoclonal Antibody [clone: EP394Y]
Supplier: Abcam
AB33886-100 AB33886-1ML
ABCAAB33886-100EA 613 EUR
ABCAAB33886-100 ABCAAB33886-1ML
Anti-General Receptor for phosphoinositides 1/GRASP Rabbit Monoclonal Antibody [clone: EP394Y]
Antibodies
Caution:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: General Receptor for phosphoinositides 1
Clonality: monoclonal
Clone: EP394Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now