Anti-Junctional Adhesion Molecule 1/JAM-A Rabbit Monoclonal Antibody [clone: EP1042Y]

Supplier: Abcam
AB52647-100 AB52647-1ML
ABCAAB52647-100EA 694 EUR
ABCAAB52647-100 ABCAAB52647-1ML
Anti-Junctional Adhesion Molecule 1/JAM-A Rabbit Monoclonal Antibody [clone: EP1042Y]
Antibodies
Rabbit monoclonal [EP1042Y] to Junctional Adhesion Molecule 1/JAM-A.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Junctional Adhesion Molecule 1/JAM-A
Clonality: Monoclonal
Clone: EP1042Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR