Anti-BRAF (phospho T401) Rabbit Monoclonal Antibody [clone: EPR2208Y]

Supplier: Abcam
AB68215-100 AB68215-20UL AB68215-1ML
ABCAAB68215-100EA 682 EUR
ABCAAB68215-100 ABCAAB68215-20UL ABCAAB68215-1ML
Anti-BRAF (phospho T401) Rabbit Monoclonal Antibody [clone: EPR2208Y]
Antibodies
Rabbit monoclonal [EPR2208Y] to BRAF (phospho T401).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BRAF
Clonality: Monoclonal
Clone: EPR2208Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR