Anti-CD8 alpha Rabbit Monoclonal Antibody [clone: SP16]

Supplier: Abcam
AB101500-100 AB101500-1 AB101500-500
ABCAAB101500-100EA 807 EUR
ABCAAB101500-100 ABCAAB101500-1 ABCAAB101500-500
Anti-CD8 alpha Rabbit Monoclonal Antibody [clone: SP16]
Antibodies
Rabbit monoclonal [SP16] to CD8 alpha.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD8 alpha
Clonality: Monoclonal
Clone: SP16
Conjugation:
Epitope: C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR