Anti-Cytokeratin 17 Rabbit Monoclonal Antibody [clone: EP1623]

Supplier: Abcam
AB109725-100 AB109725-1ML
ABCAAB109725-100EA 475 EUR
ABCAAB109725-100 ABCAAB109725-1ML
Anti-Cytokeratin 17 Rabbit Monoclonal Antibody [clone: EP1623]
Antibodies
Rabbit monoclonal [EP1623] to Cytokeratin 17 - Cytoskeleton Marker.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cytokeratin 17
Clonality: Monoclonal
Clone: EP1623
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR