Anti-IgG Donkey Polyclonal Antibody (CF™ 633)

Supplier: Biotium
20076 20076-1
20076.EA 359 EUR
20076. 20076-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR