Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Supplier: Biotium
20084 20084-1
20084.EA 315 EUR
20084. 20084-1
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antibodies
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR