Anti-IgG Donkey Polyclonal Antibody (Biotin)

Supplier: Biotium
20189 20189-1
20189.EA 359 EUR
20189. 20189-1
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR