Anti-IgG Donkey Polyclonal Antibody (CF™ 640R)
Supplier: Biotium
20199 20199-1
20199.EA 359 EUR
20199. 20199-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 640R)
Antibodies
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rat