Anti-IgG Rabbit Polyclonal Antibody (CF™ 640R)

Supplier: Biotium
20200 20200-1
20200.EA 315 EUR
20200. 20200-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 640R)
Antibodies
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR