Anti-CRYAB Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
NB100-2519 NB100-2519SS
NOVUNB100-2519EA 623 EUR
NOVUNB100-2519 NOVUNB100-2519SS
Anti-CRYAB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse