Anti-XAGE1A Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
NB100-262 NB100-262SS
NOVUNB100-262EA 566 EUR
NOVUNB100-262 NOVUNB100-262SS
Anti-XAGE1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: XAGE1A (X antigen family, member 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human