Anti-LIPE Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-37253 NB110-37253SS
NOVUNB110-37253EA 492 EUR
NOVUNB110-37253 NOVUNB110-37253SS
Anti-LIPE Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LIPE (lipase, hormone-sensitive)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR