Anti-GPIHBP1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-41539 NB110-41539SS
NOVUNB110-41539EA 591 EUR
NOVUNB110-41539 NOVUNB110-41539SS
Anti-GPIHBP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPIHBP1 (glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR