Anti-CITED4 Mouse monoclonal antibody unconjugated [clone: HT13-2D6.3]

Supplier: Novus Biologicals
NB110-41572 NB110-41572SS
NOVUNB110-41572EA 554 EUR
NOVUNB110-41572 NOVUNB110-41572SS
Anti-CITED4 Mouse monoclonal antibody unconjugated [clone: HT13-2D6.3]
Antibodies
Type: Primary
Antigen: CITED4 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4)
Clonality: monoclonal
Clone: HT13-2D6.3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR