Anti-ATG5 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
NB110-53818 NB110-53818SS
NOVUNB110-53818EA 735 EUR
NOVUNB110-53818 NOVUNB110-53818SS
Anti-ATG5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATG5 (autophagy related 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now