Anti-TP53 Mouse Monoclonal Antibody [clone: PAb 240]

Supplier: Novus Biologicals
NB200-103 NB200-103SS
NOVUNB200-103EA 654 EUR
NOVUNB200-103 NOVUNB200-103SS
Anti-TP53 Mouse Monoclonal Antibody [clone: PAb 240]
Antibodies
The p53 Antibody (PAb 240) from Novus Biologicals is a mouse monoclonal antibody to p53. This antibody reacts with human, mouse, rat, yeast, xenopus (negative). The p53 Antibody (PAb 240) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.

Recommended Dilutions: Immunohistochemistry (Parafin): 1/250 - 1/500

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: PAb 240
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR