Anti-MYCN Mouse Monoclonal Antibody [clone: NMYC-1]

Supplier: Novus Biologicals
NB200-109 NB200-109SS
NOVUNB200-109EA 630 EUR
NOVUNB200-109 NOVUNB200-109SS
Anti-MYCN Mouse Monoclonal Antibody [clone: NMYC-1]
Antibodies
Type: Primary
Antigen: MYCN (v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog)
Clonality: monoclonal
Clone: NMYC-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR