Anti-BIRC5 Mouse Monoclonal Antibody [clone: 8E2]
Supplier: Novus Biologicals
NB500-644 NB500-644SS
NOVUNB500-644EA 567 EUR
NOVUNB500-644 NOVUNB500-644SS
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 8E2]
Antibodies
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: 800
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat