Anti-ATF3 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
NBP1-02935 NBP1-02935SS
NOVUNBP1-02935EA 616 EUR
NOVUNBP1-02935 NOVUNBP1-02935SS
Anti-ATF3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATF3 (activating transcription factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now