Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: MIC1-1C3]

Supplier: Novus Biologicals
NBP1-18961 NBP1-18961SS
NOVUNBP1-18961EA 648 EUR
NOVUNBP1-18961 NOVUNBP1-18961SS
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: MIC1-1C3]
Antibodies
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: MIC1-1C3
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR