Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-2A6]
Supplier: Novus Biologicals
NBP1-18967 NBP1-18967SS
NOVUNBP1-18967EA 554 EUR
NOVUNBP1-18967 NOVUNBP1-18967SS
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-2A6]
Antibodies
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-2A6
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse